Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Chloride-Ammonium Hydroxide TS, (U.S.P. Test Solution), Spectrum™

Click to view available options
Quantity:
100 mL
1 L
Packaging:
Amber Glass Bottle
7664-41-7,12125-02-9
H3N
17.03
QGZKDVFQNNGYKY-UHFFFAOYSA-N
ammonia
N
7664-41-7,12125-02-9
H3N
17.03
QGZKDVFQNNGYKY-UHFFFAOYSA-N
ammonia
N

A-323, 12125-02-9

Spectrum™ Ammonium Chloride-Ammonium Hydroxide TS, (U.S.P. Test Solution) is a solution, sometimes designated with "TS or Test Solution", that is prepared according to the specifications of the USP Pharmacopeial Convention, for the purpose of conducting USP and National Formulary tests and assays.
CAS Max % 0.001,0.999
UN Number UN2672
Packaging Amber Glass Bottle
Quantity 1 L