Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ammonium Chloride (Crystalline/Certified ACS), Fisher Chemical™
Click to view available options
Quantity:
500 g
3 kg
10 kg
Packaging:
Poly Bottle
Poly Pail
Specifications
Melting Point | 340°C |
Color | White |
pH | 5.0 |
Physical Form | Solid |
Quantity | 3 kg |
Assay Percent Range | ≥99.5% |
Formula Weight | 53.49 |
Percent Purity | ≥99.5% |
Grade | Certified ACS |
Identification | Pass Test |
Show More |
Safety and Handling
Hazard Statement | WARNING! |
Precautionary Statement | Emergency Overview Harmful if swallowed. May cause skin and respiratory tract irritation. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Get medical attention immediately if symptoms occur. NFPA Health:2 Flammability:0 Instability:1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction