Ammonium Chloride-EDTA Solution, for Nitrate by the Cadmium Reduction Method, Ricca Chemical

Manufacturer: Ricca Chemical CompanySupplier Diversity Partner 62616

Packaging Poly Natural
Quantity 500mL

 View more versions of this product

Catalog No. 626-16

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12125-02-9 (Ammonium Chloride),1336-21-6 (Ammonium Hydroxide),7732-18-5 (Water, Deionized)
    Physical Form Liquid
    Chemical Name or Material Ammonium Chloride-EDTA Solution

    Specifications

    Color Clear colorless
    Physical Form Liquid
    Quantity 500mL
    CAS 12125-02-9 (Ammonium Chloride),1336-21-6 (Ammonium Hydroxide),7732-18-5 (Water, Deionized)
    Packaging Poly Natural
    Chemical Name or Material Ammonium Chloride-EDTA Solution

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit