Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Chloride, Granular, USP, 99.5-100.5%, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
12125-02-9
ClH4N
53.49
NLXLAEXVIDQMFP-UHFFFAOYSA-N
ammonium chloride
[NH4+].[Cl-]
12125-02-9
ClH4N
53.49
NLXLAEXVIDQMFP-UHFFFAOYSA-N
ammonium chloride
[NH4+].[Cl-]

AM175, 12125-02-9, NH4Cl

Spectrum™ Ammonium Chloride, Granular, USP is used as an expectorant in cough medicine. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 1
Percent Purity 99.5 to 100.5%
Ignition Residue 0.001
Loss on Drying 0.005
Packaging Poly Bottle
pH Range 4.6-6.0
Quantity 2.5 kg