Ammonium chloride, Puratronic, 99.999% (metals basis), Alfa Aesar

Manufacturer: Alfa Aesar 1063230

Quantity 250g

 View more versions of this product

Catalog No. AA1063230

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12125-02-9
    Physical Form Powder
    Assay Percent Range 100.00%
    Molecular Formula NH4Cl
    Formula Weight 53.49
    MDL Number MFCD00011420
    Chemical Name or Material Ammonium chloride

    Specifications

    Formula Weight 53.49
    Quantity 250g
    Color White
    EINECS Number 235-186-4
    Physical Form Powder
    Molecular Formula NH4Cl
    Odor Odorless
    Assay Percent Range 100.00%
    Appearance White powder
    MDL Number MFCD00011420
    CAS 12125-02-9
    Health Hazard 1 H302-H319
    Storage Note 1 Hygroscopic
    Chemical Name or Material Ammonium chloride

    Hygroscopic

    H302-H319

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit