Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Chloride TS, (U.S.P. Test Solution), Spectrum™

Click to view available options
Quantity:
500 mL
Packaging:
Poly Bottle

A-362, 12125-02-9

Spectrum™ Ammonium Chloride TS, (U.S.P. Test Solution) is a solution, sometimes designated with "TS or Test Solution", that is prepared according to the specifications of the USP Pharmacopeial Convention, for the purpose of conducting USP and National Formulary tests and assays.
CAS 12125-02-9,7732-18-5
CAS Max % 0.001,0.999
Molecular Formula ClH4N
InChI Key NLXLAEXVIDQMFP-UHFFFAOYSA-N
SMILES [NH4+].[Cl-]
IUPAC Name ammonium chloride
Molecular Weight (g/mol) 53.49
Packaging Poly Bottle
Quantity 500 mL