Ammonium chromate, Reagent Grade, Alfa Aesar™

Manufacturer: Alfa Aesar 8768022

Quantity 100g

 View more versions of this product

Catalog No. AA8768022

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 7788-98-9
    Physical Form Crystalline
    Molecular Formula (NH4)2CrO4
    Formula Weight 152.07
    MDL Number MFCD00015958
    Chemical Name or Material Ammonium chromate
    Grade Reagent Grade

    Specifications

    Formula Weight 152.07
    Quantity 100g
    Color Yellow-orange
    DOT Information DOT Class: 5.1, Packaging Group: III
    Density 1.91g/cm3
    EINECS Number 232-138-4
    Physical Form Crystalline
    Odor Odorless
    Molecular Formula (NH4)2CrO4
    Appearance Yellow-orange crystalline
    MDL Number MFCD00015958
    CAS 7788-98-9
    Health Hazard 1 H350-H400-H410-H317
    Chemical Name or Material Ammonium chromate
    Grade Reagent Grade

    H350-H400-H410-H317

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit