Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Citrate Dibasic, ACS, 98-103%, Spectrum™

Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
3012-65-5
3012-65-5
Grade ACS
CAS Max % 1
Percent Purity 98 to 103%
Chloride 1e-05
Heavy Metals 5 ppm
Insoluble Matter 5e-05
Iron (Fe) 1e-05
Packaging Fiber Drum
Phosphate 5 ppm
Residue after Ignition 0.0001
Sulfur Compounds 5e-05
Quantity 45 kg
Show More Show Less

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.