Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Citrate Dibasic (Crystalline/Certified ACS), Fisher Chemical™

Catalog Number A663212
Click to view available options
Quantity:
500 g
2.5 kg
3012-65-5
C6H14N2O7
226.19
MFCD00013068
YXVFQADLFFNVDS-UHFFFAOYSA-N
ammonium citrate dibasic, ammonium hydrogencitrate, diammonium hydrogen citrate, citric acid diammonium salt, citric acid ammonium salt, di-ammonium hydrogen citrate, citro diamine, citric acid diamine, ammonium citrate, dibasic, acs, ammonium citrate dibasic, puriss
13710713
CHEBI:63076
diammonium 3-carboxy-3-hydroxypentanedioate
[NH4+].[NH4+].OC(=O)C(O)(CC([O-])=O)CC([O-])=O
3012-65-5
C6H14N2O7
226.19
MFCD00013068
YXVFQADLFFNVDS-UHFFFAOYSA-N
ammonium citrate dibasic, ammonium hydrogencitrate, diammonium hydrogen citrate, citric acid diammonium salt, citric acid ammonium salt, di-ammonium hydrogen citrate, citro diamine, citric acid diamine, ammonium citrate, dibasic, acs, ammonium citrate dibasic, puriss
13710713
CHEBI:63076
diammonium 3-carboxy-3-hydroxypentanedioate
[NH4+].[NH4+].OC(=O)C(O)(CC([O-])=O)CC([O-])=O
Color White
pH 4.8 to 5.3
Physical Form Solid
Quantity 2.5 kg
Assay Percent Range 98.0 to 103.0%
Linear Formula (NH4)2O2CCH2C(OH)(CO2H)CH2CO2
Formula Weight 226.19
Percent Purity 98.0 to 103.0%
Grade Certified ACS
Identification Pass Test
Packaging Poly Pail
Density 1.5g/cm³
Chemical Name or Material Ammonium Citrate Dibasic
Show More Show Less
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit