Ammonium Citrate Dibasic (Crystalline/Certified ACS), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.

Quantity 500g

 View more versions of this product

Catalog No. A663-500

  Check Availability
Add to Cart

Custom packing/Bulk ordering

You can now order this item in custom quantities.  Learn More

    Chemical Identifiers

    CAS 3012-65-5
    Physical Form Solid
    Assay Percent Range 98.0 to 103.0%
    Molecular Formula (NH4)2C6H6O7
    Linear Formula (NH4)2O2CCH2C(OH)(CO2H)CH2CO2
    Formula Weight 226.19
    MDL Number MFCD00013068
    Synonym Citric Acid Diammonium Salt
    Chemical Name or Material Ammonium Citrate Dibasic
    Assay 98.0 to 103.0%
    Grade Certified ACS Grade

    Specifications

    Formula Weight 226.19
    Quantity 500g
    Assay Percent Range 98.0 to 103.0%
    Viscosity [NULL]
    Density 1.5g/cm³
    Physical Form Solid
    Color White
    pH 4.8 to 5.3
    Chloride 0.001% max.
    Heavy Metals (as Pb) 5ppm max.
    Identification Pass Test
    Ignition Residue 0.01% max.
    Insoluble Matter 0.005% max.
    Iron (Fe) 0.001% max.
    Oxalate P.T. (about 0.05%)
    Packaging Glass Bottle
    Phosphate 5ppm max.
    Molecular Formula (NH4)2C6H6O7
    Linear Formula (NH4)2O2CCH2C(OH)(CO2H)CH2CO2
    Sulfur Compounds 0.005% max.
    MDL Number MFCD00013068
    CAS 3012-65-5
    ChemAlert Storage Symbol Gray
    Synonym Citric Acid Diammonium Salt
    Chemical Name or Material Ammonium Citrate Dibasic
    Assay 98.0 to 103.0%
    Grade Certified ACS Grade
     

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling
  • Product Certifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit