Alfa Aesar™ Ammonium-d4 deuteroxide, 99% (Isotopic), 23.0-27.0 wt% soln. in D2O

Manufacturer: Alfa Aesar™ 2298006

Quantity 5g

 View more versions of this product

Catalog No. AA2298006

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12168-30-8
    Linear Formula ND4OD
    Formula Weight 40.09
    MDL Number MFCD00064511
    UN Number UN2672
    Synonym Ammonium hydroxide, deuterated
    Chemical Name or Material Ammonium-d4 deuteroxide, Isotopic

    Specifications

    Formula Weight 40.09
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Physical Form Liquid
    MDL Number MFCD00064511
    CAS 12168-30-8
    Synonym Ammonium hydroxide, deuterated
    Health Hazard 1 Danger
    Sensitivity Moisture Sensitive
    Quantity 5g
    Color Colorless
    Linear Formula ND4OD
    Refractive Index 1.3340
    UN Number UN2672
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMMONIA SOLUTION
    Concentration 23.0 to 27.0 wt% soln. in D2O
    Chemical Name or Material Ammonium-d4 deuteroxide, Isotopic
    Assay 99%

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling
  • Promotions

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit