Learn More
Ammonium dichromate, 99%, ACROS Organics™
Description
Specifications
Assay Percent Range | 98.5% min. (Iodometry) |
Linear Formula | (NH4)2Cr2O7 |
Merck Index | 15, 512 |
Solubility Information | Solubility in water: 360g/L (20°C). Other solubilities: soluble in alcohol |
Formula Weight | 252.07 |
Physical Form | Adhering Crystals |
Percent Purity | 99% |
Chloride | 500ppm max |
Chlorine (Cl) | 500ppm max. |
Loss on Drying | 3% max. (105°C, 4 hrs) |
Show More |
Safety and Handling
GHS H Statement
Harmful in contact with skin.
Very toxic to aquatic life with long lasting effects.
May damage fertility.
May damage the unborn child.
May cause genetic defects.
Causes damage to organs through prolonge
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. IF ON SKIN: Wash with plenty of soap and water.
GHS Signal Word: Danger
EINECSNumber : 232-143-1
RTECSNumber : HX7650000
TSCA : TSCA