Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Dichromate, Crystal, 99.5%, Spectrum™

Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
7789-09-5
7789-09-5
Grade Reagent
CAS Max % 1
Percent Purity 0.995
Calcium (Ca) 5e-05
Chloride 5e-05
Insoluble Matter 5e-05
Iron (Fe) 5e-05
Packaging Fiber Drum
Sulfate 0.0001
pH 3 to 5
Quantity 50 kg
Show More Show Less

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.