Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Dichromate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
2.5kg
500g
Packaging:
Poly Bottle

Chemical Identifiers

7789-09-5
Cr2H8N2O7
252.06
JOSWYUNQBRPBDN-UHFFFAOYSA-P
24600

High quality chemicals for laboratory and specialized industrial use

CAS 7789-09-5
Molecular Formula Cr2H8N2O7
InChI Key JOSWYUNQBRPBDN-UHFFFAOYSA-P
SMILES [NH4+].[NH4+].[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-]
IUPAC Name diazanium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium
Molecular Weight (g/mol) 252.06
PubChem CID 24600
Formula Weight 252.07
Physical Form Crystals
Packaging Poly Bottle
Specific Gravity 1L = 2.15kg
Quantity 2.5kg
Chemical Name or Material Ammonium Dichromate
Grade Baker Analyzed™ ACS Reagent
Show More Show Less