Ammonium Dichromate (Crystalline/Certified), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.

Packaging Glass Bottle
Quantity 100g

 View more versions of this product

Catalog No. A644-100

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 

Custom packing/Bulk ordering

You can now order this item in custom quantities.  Learn More

    Chemical Identifiers

    CAS 7789-09-5
    Physical Form Solid
    Molecular Formula H8Cr2N2O7
    Linear Formula (NH4)2Cr2O7
    Formula Weight 252.07
    MDL Number MFCD00010879
    Synonym Chromic Acid Diammonium Salt
    Chemical Name or Material Ammonium Dichromate
    Grade Certified Grade

    Specifications

    Melting Point 170°C
    Formula Weight 252.07
    Quantity 100g
    DOT Information DOT Class 5.1, : Oxidizer
    Viscosity [NULL]
    Physical Form Solid
    Color Orange
    pH 3 to 4
    Density 2.15g/mL
    Molecular Formula H8Cr2N2O7
    Identification Pass Test
    Packaging Glass Bottle
    Calcium (Ca) 0.002% max.
    Chloride 0.005% max.
    Insoluble Matter 0.005% max.
    Alkalis and Other Salts 0.15% max.
    Linear Formula (NH4)2Cr2O7
    Sulfate 0.006% max.
    MDL Number MFCD00010879
    CAS 7789-09-5
    ChemAlert Storage Symbol Yellow
    Synonym Chromic Acid Diammonium Salt
    Chemical Name or Material Ammonium Dichromate
    Grade Certified Grade
     

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling
  • Product Certifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit