Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Dichromate (Crystalline/Certified), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.

Packaging Glass Bottle
Quantity 2.5kg

 View more versions of this product

Catalog No. A644-212


 

Due to product restrictions, please to purchase or view availability for this product.

Custom packing/Bulk ordering

You can now order this item in custom quantities. Learn More

Specifications Description & Specifications

Chemical Identifiers

7789-09-5
H8Cr2N2O7
252.07
Chromic Acid Diammonium Salt
Certified Grade
Solid
(NH4)2Cr2O7
MFCD00010879
Ammonium Dichromate

Specifications

7789-09-5
Solid
(NH4)2Cr2O7
MFCD00010879
0.002% max.
2.15g/mL
0.005% max.
0.006% max.
170°C
2.5kg
Certified Grade
0.15% max.
H8Cr2N2O7
252.07
Chromic Acid Diammonium Salt
0.005% max.
Pass Test
Glass Bottle
Orange
3 to 4
Ammonium Dichromate

Safety and Handling

SDS & Safety Documents

DOTInformation : DOT Class 5.1, : Oxidizer

Alternative Products
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit