Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Dichromate, Honeywell

Click to view available options
Quantity:
100g
500g
1kg
7789-09-5
Cr2H8N2O7
252.06
MFCD00010879
JOSWYUNQBRPBDN-UHFFFAOYSA-P
ammonium dichromate,ammonium bichromate,diammonium dichromate,ammonium dichromate vi,ammoniumbichromaat dutch,ammoniumdichromaat dutch,ammoniumdichromat german,unii-5j18bp595g,hsdb 481
24600
diazanium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium
[NH4+].[NH4+].[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-]
7789-09-5
Cr2H8N2O7
252.06
MFCD00010879
JOSWYUNQBRPBDN-UHFFFAOYSA-P
ammonium dichromate,ammonium bichromate,diammonium dichromate,ammonium dichromate vi,ammoniumbichromaat dutch,ammoniumdichromaat dutch,ammoniumdichromat german,unii-5j18bp595g,hsdb 481
24600
diazanium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium
[NH4+].[NH4+].[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-]

Purum p.a.

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
Linear Formula (NH4)2Cr2O7
UN Number 1439
Formula Weight 252.06g/mol
Percent Purity ≥97% (RT)
Grade purum p.a.
Vapor Density 8.7 (vs air)
Quantity 100g
Chemical Name or Material Ammonium Dichromate

P201-P280-P304 + P340 + P310-P305 + P351 + P338-P308 + P313

H272-H301-H312-H314-H317-H330-H334-H340-H350-H360FD-H372-H410

EINECSNumber : 232-143-1