Alfa Aesar™ Ammonium dihydrogen phosphate, 98%

Manufacturer: Alfa Aesar™ A1528336

Quantity 500g

 View more versions of this product

Catalog No. AAA1528336

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7722-76-1
    Molecular Formula NH4H2PO4
    Formula Weight 115.03
    MDL Number MFCD00003396
    Merck Index 14,543
    Synonym mono-Ammonium phosphate; Ammonium phosphate, monobasic
    Chemical Name or Material Ammonium dihydrogen phosphate

    Specifications

    Formula Weight 115.03
    TSCA TSCA
    MDL Number MFCD00003396
    Merck Index 14,543
    Melting Point 198° to 204°C
    Quantity 500g
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Odor Odorless
    Molecular Formula NH4H2PO4
    CAS 7722-76-1
    Synonym mono-Ammonium phosphate; Ammonium phosphate, monobasic
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Chemical Name or Material Ammonium dihydrogen phosphate
    Assay 98%

    Description

    Applications
    Ammonium dihydrogen phosphate is widely utilized as a fertilizer, which provides phosphorous and nitrogen for plants. It is used as a dry fire extinguisher. It is a well known nonlinear optical material for various optoelectronic applications. It is also used in an emerald green, amethyst and aquamarine crystal growing box kit for children.

    Solubility
    Soluble in water. Slightly soluble in alcohol. Insoluble in acetone.

    Notes
    Incompatible with strong oxidizing agents, strong acids and sodium hypochlorite.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit