Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium dihydrogen phosphate, 99.9%, (trace metal basis), Acros Organics

Click to view available options
Quantity:
100g
500g
2.5kg
Packaging:
Plastic bottle
7722-76-1
H6NO4P
115.03
LFVGISIMTYGQHF-UHFFFAOYSA-N
ammonium dihydrogen phosphate,monoammonium phosphate,ammonium biphosphate,ammonium acid phosphate,phosphoric acid, monoammonium salt,ammonium monophosphate,ammonium dihydrogen orthophosphate,ammonium dihydrophosphate,ammonium diacid phosphate,primary ammonium phosphate
24402
CHEBI:62982
azanium;dihydrogen phosphate
[NH4+].OP(=O)(O)[O-]
7722-76-1
H6NO4P
115.03
LFVGISIMTYGQHF-UHFFFAOYSA-N
ammonium dihydrogen phosphate,monoammonium phosphate,ammonium biphosphate,ammonium acid phosphate,phosphoric acid, monoammonium salt,ammonium monophosphate,ammonium dihydrogen orthophosphate,ammonium dihydrophosphate,ammonium diacid phosphate,primary ammonium phosphate
24402
CHEBI:62982
azanium;dihydrogen phosphate
[NH4+].OP(=O)(O)[O-]
Assay Percent Range 99.90%
Linear Formula NH4H2PO4
Merck Index 15,541
Solubility Information Solubility in water: 368g/L (20°C). Other solubilities: slightly soluble in alcohol,practically insoluble in acetone
Formula Weight 115.03
Physical Form Crystalline Powder
Percent Purity 99.9%
Grade Trace Metal Basis
Packaging Plastic bottle
Solubility Solubility in water: 368g/L (20°C). Other solubilities: slightly soluble in alcohol, practically insoluble in acetone
Total Trace Metal Impurities 0.1% max.
Color White
Melting Point 190.0°C
Quantity 500g
Chemical Name or Material Ammonium dihydrogen phosphate
Show More Show Less

EINECSNumber : 231-764-5