Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium dihydrogen phosphate, 99+%, for analysis, ACROS Organics™

Click to view available options
Quantity:
250g
1kg
2.5kg
10kg
Packaging:
Plastic Drum
Plastic bottle
7722-76-1
H6NO4P
115.03
MFCD00003396
LFVGISIMTYGQHF-UHFFFAOYSA-N
ammonium dihydrogen phosphate,monoammonium phosphate,ammonium biphosphate,ammonium acid phosphate,phosphoric acid, monoammonium salt,ammonium monophosphate,ammonium dihydrogen orthophosphate,ammonium dihydrophosphate,ammonium diacid phosphate,primary ammonium phosphate
24402
CHEBI:62982
azanium;dihydrogen phosphate
[NH4+].OP(=O)(O)[O-]
7722-76-1
H6NO4P
115.03
MFCD00003396
LFVGISIMTYGQHF-UHFFFAOYSA-N
ammonium dihydrogen phosphate,monoammonium phosphate,ammonium biphosphate,ammonium acid phosphate,phosphoric acid, monoammonium salt,ammonium monophosphate,ammonium dihydrogen orthophosphate,ammonium dihydrophosphate,ammonium diacid phosphate,primary ammonium phosphate
24402
CHEBI:62982
azanium;dihydrogen phosphate
[NH4+].OP(=O)(O)[O-]
Assay Percent Range 99+%
Linear Formula NH4H2PO4
Merck Index 15,541
Arsenic (As) 0.5ppm max.
Solubility Information Solubility in water: 368g/L (20°C). Other solubilities: slightly soluble in alcohol,practically insoluble in acetone
Formula Weight 115.03
Physical Form Crystalline Solid
Percent Purity 99+%
Grade Analysis
Chloride 5ppm max
Chlorine (Cl) 5ppm max.
Density 1.8g/mL
Heavy Metals (as Pb) 5ppm max.
Iron (Fe) 10ppm max.
Nitrate 10ppm max.
Packaging Plastic bottle
Potassium (K) 50ppm max.
Sodium (Na) 50ppm max.
Solubility Solubility in water: 368g/L (20°C). Other solubilities: slightly soluble in alcohol, practically insoluble in acetone
Sulfate 50ppm max.
Color White
Melting Point 190.0°C
pH 4 to 4.5 (5% soln. at 20°C)
Quantity 2.5kg
Chemical Name or Material Ammonium dihydrogen phosphate
Show More Show Less

EINECSNumber : 231-764-5