Alfa Aesar™ Ammonium dihydrogen phosphate& Magnesium nitrate, Matrix Modifier Solution, Specpure™

Manufacturer: Alfa Aesar™ 45292AK

Quantity 250mL

 View more versions of this product

Catalog No. AA45292AK

  Check Availability
Add to Cart

    Chemical Identifiers

    MDL Number MFCD09910542
    UN Number UN3264
    Chemical Name or Material Ammonium dihydrogen phosphate & Magnesium nitrate Matrix Modifier Solution
    Grade Specpure™

    Specifications

    Health Hazard 3 GHS P Statement
    P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Take off contaminated clothing and wash before reuse.
    Specific treatment (see label).
    If skin irritation occurs: Get medical advice/attention.
    If eye irritation persists: Get medical advice/attention.
    Physical Form Liquid
    Concentration 0.001% NH4H2PO4 & 0.06% Mg(NO3)2 in 2% HNO3
    Quantity 250mL
    TSCA TSCA
    MDL Number MFCD09910542
    UN Number UN3264
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319
    Causes skin irritation.
    Causes serious eye irritation.
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.
    Chemical Name or Material Ammonium dihydrogen phosphate & Magnesium nitrate Matrix Modifier Solution
    Grade Specpure™

    GHS H Statement
    H315-H319
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Take off contaminated clothing and wash before reuse.
    Specific treatment (see label).
    If skin irritation occurs: Get medical advice/attention.
    If eye irritation persists: Get medical advice/attention.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit