Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Fluoborate, Crystal, Technical, Spectrum™

Click to view available options
Quantity:
125 g
500 g
2.5 kg
50 lb.
Packaging:
Poly Bottle
Poly Pail
13826-83-0
13826-83-0
Grade Technical
CAS Max % 1
Packaging Poly Bottle
Quantity 500 g

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.