Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ammonium fluoride, 98+%, extra pure, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic Drum
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Colorless or White |
Physical Form | Adhering Crystals and/or Chunks |
Quantity | 2.5 kg |
Assay Percent Range | 98+% |
Linear Formula | NH4F |
Merck Index | 15, 519 |
Solubility Information | Solubility in water: soluble. Other solubilities: slightly soluble in alcohol |
Formula Weight | 37.04 |
Percent Purity | ≥98% |
Grade | Extra Pure |
Show More |
Safety and Handling
Hazard Category | GHS Signal Word: Danger |
Hazard Statement | GHS H Statement Toxic in contact with skin. Toxic if inhaled. Toxic if swallowed. |
Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. Do not breathe dust/fume/gas/mist/vapors/spray. IF ON SKIN: Gently wash with plenty of soap and water. IF INHALED: Remove victim to fresh air |
EINECSNumber | 235-185-9 |
RTECSNumber | BQ6300000 |
TSCA | TSCA |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction