Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium fluoride, 98+%, extra pure, Thermo Scientific Chemicals

Catalog Number AC201841000
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic Drum
Plastic bottle
12125-01-8
H4FN
37.04
MFCD00011423
12125-01-8
H4FN
37.04
MFCD00011423

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless or White
Physical Form Adhering Crystals and/or Chunks
Quantity 100 g
Assay Percent Range 98+%
Linear Formula NH4F
Merck Index 15, 519
Solubility Information Solubility in water: soluble. Other solubilities: slightly soluble in alcohol
Formula Weight 37.04
Percent Purity ≥98%
Grade Extra Pure
Packaging Plastic bottle
Residue after Ignition 0.01 % max. (SO4)
Chemical Name or Material Ammonium fluoride, p.a.
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic in contact with skin.
Toxic if inhaled.
Toxic if swallowed.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air
EINECSNumber 235-185-9
RTECSNumber BQ6300000
TSCA TSCA

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit