Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Fluoride, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
2.5kg
500g
Packaging:
Poly Bottle
Poly Pail

Chemical Identifiers

12125-01-8
NH4F

High quality chemicals for laboratory and specialized industrial use

CAS 12125-01-8
Molecular Formula NH4F
Formula Weight 37.04
Physical Form Crystals
Packaging Poly Bottle
Quantity 500g
Chemical Name or Material Ammonium Fluoride
Grade Baker Analyzed™ ACS Reagent