Alfa Aesar™ Ammonium formate, 97%, water <3%

Manufacturer: Alfa Aesar™ A1069930

Quantity 250g

 View more versions of this product

Catalog No. AAA1069930

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 540-69-2
    Molecular Formula CH5NO2
    Linear Formula HCO2NH4
    Formula Weight 63.06
    MDL Number MFCD00013103
    Beilstein 3625095
    Merck Index 14,523
    Synonym Formic acid ammonium salt
    Chemical Name or Material Ammonium formate, Water <3%

    Specifications

    Formula Weight 63.06
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    MDL Number MFCD00013103
    CAS 540-69-2
    Merck Index 14,523
    Melting Point ≈116°C
    Synonym Formic acid ammonium salt
    RTECS Number BQ6650000
    Molecular Formula CH5NO2
    Linear Formula HCO2NH4
    Quantity 250g
    Beilstein 3625095
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Odor Ammonia-like
    Health Hazard 1 Warning
    Chemical Name or Material Ammonium formate, Water <3%
    Assay 97%

    Description

    Applications
    Ammonium formate is widely used in various organic reactions like Leuckart reaction which involves the reductive amination of aldehydes and ketones. It serves as a buffer in high performance liquid chromatography (HPLC) and liquid chromatography-mass spectrometry (LC/MS). It finds application in palladium on carbon (Pd/C) reduction of functional groups. For example, reduction of alkenes to alkanes and formaldehyde to methanol. It is also used to prepare formic acid insitu as well as used to store formic acid by making it as an ammonium salt.

    Solubility
    Soluble in water, liquid ammonia and alcohol.

    Notes
    Hygroscopic. Incompatible with strong acids and strong oxidizing agents.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit