Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ Ammonium Formate, Reagent grade, 97%, Honeywell™ Burdick & Jackson™

Click to view available options
Quantity:
100g
500g
1kg
2.5kg
Packaging:
Poly Bottle
540-69-2
CH5NO2
63.056
MFCD00013103
VZTDIZULWFCMLS-UHFFFAOYSA-N
ammonium formate,formic acid, ammonium salt,formic acid ammonium salt,ammoniumformate,azanium formate,ammonium formiate,formic acid, ammonium salt 1:1,mravencan amonny czech,hsdb 479,mravencan amonny
2723923
CHEBI:63050
azanium;formate
C(=O)[O-].[NH4+]
540-69-2
CH5NO2
63.056
MFCD00013103
VZTDIZULWFCMLS-UHFFFAOYSA-N
ammonium formate,formic acid, ammonium salt,formic acid ammonium salt,ammoniumformate,azanium formate,ammonium formiate,formic acid, ammonium salt 1:1,mravencan amonny czech,hsdb 479,mravencan amonny
2723923
CHEBI:63050
azanium;formate
C(=O)[O-].[NH4+]

Reagent Grade, 97%

  • ≥95% for general laboratory use.
Linear Formula HCO2NH4
UN Number NONH for all modes of transport
Beilstein 3625095
Appearance Crystals/Granules
Formula Weight 63.06g/mol
Percent Purity 97%
Grade Reagent
Density 1.26g/mL at 25°C
Packaging Poly Bottle
Color White
Melting Point 116°C
pH 6.0 to 7.0 (at 20°C)
Quantity 1kg
Chemical Name or Material Ammonium Formate
Show More Show Less

EINECSNumber : 208-753-9

RTECSNumber : BQ6650000