Alfa Aesar™ Ammonium hexabromoosmate(IV), 99.9% (metals basis), Os 26.5 % min

Manufacturer: Alfa Aesar™ 1264803

Quantity 1g

 View more versions of this product

Catalog No. AA1264803

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 24598-62-7
    Formula Weight 705.73
    MDL Number MFCD00049389
    Chemical Name or Material Ammonium hexabromoosmate(IV)

    Specifications

    Formula Weight 705.73
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00049389
    Quantity 1g
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H302-H315-H319-H335
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Concentration Os 26.5 % min.
    Physical Form Powder
    Color Black
    TSCA TSCA
    Odor Odorless
    Sensitivity Hygroscopic
    CAS 24598-62-7
    Chemical Name or Material Ammonium hexabromoosmate(IV)
    Assay 99.9%, (metals basis)

    GHS H Statement
    H302-H315-H319-H335
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit