Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium hexabromoplatinate(IV), 99.99%, (trace metal basis), ACROS Organics™

Click to view available options
Quantity:
1g
2g
10g
Packaging:
Glass bottle
17363-02-9
Br6H8N2Pt
710.586
MFCD00044261
YLDMEZNZYLWNNL-UHFFFAOYSA-J
diazanium;hexabromoplatinum 2-,diammonium hexabromoplatinumdiuide,platinate 2-,hexabromo-,ammonium 1:2 , oc-6-11
16211461
diazanium;hexabromoplatinum(2-)
[NH4+].[NH4+].Br[Pt-2](Br)(Br)(Br)(Br)Br
17363-02-9
Br6H8N2Pt
710.586
MFCD00044261
YLDMEZNZYLWNNL-UHFFFAOYSA-J
diazanium;hexabromoplatinum 2-,diammonium hexabromoplatinumdiuide,platinate 2-,hexabromo-,ammonium 1:2 , oc-6-11
16211461
diazanium;hexabromoplatinum(2-)
[NH4+].[NH4+].Br[Pt-2](Br)(Br)(Br)(Br)Br
For ultra trace analysis
Assay Percent Range 99.99%
Linear Formula (NH4)2PtBr6
Formula Weight 710.61
Physical Form Crystalline Powder
Grade Trace Metal Basis
Packaging Glass bottle
Total Trace Metal Impurities 0.01% max.
Color Red
Quantity 2g
Chemical Name or Material Ammonium hexabromoplatinate(IV), 99.99%

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 241-394-6

TSCA : TSCA