Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Ammonium hexachloroiridate(III) hydrate, Premion™, 99.99% (metals basis)

Manufacturer:  Alfa Aesar™ 1071303

Quantity 1g

 View more versions of this product

Catalog No. AA1071303


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

15752-05-3
MFCD00003393
Premion™
459.06 (Anhydrous)
Ammonium hexachloroiridate(III) hydrate

Specifications

15752-05-3
459.06 (Anhydrous)
Odorless
Green
Ammonium hexachloroiridate(III) hydrate
Premion™
Crystalline Powder
MFCD00003393
Hygroscopic
1g
99.99%, (metals basis)

Safety and Handling

SDS & Safety Documents

GHS H Statement
H290-H302
May be corrosive to metals.
Harmful if swallowed.

GHS P Statement
P234-P264-P270-P301+P312-P406-P501a
Keep only in original container.
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Store in corrosive resistant container with a resistant inner liner.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit