Alfa Aesar™ Ammonium hexachloroiridate(IV), Premion™, 99.994% (metals basis), Ir 41% min

Manufacturer: Alfa Aesar™ 1071203

Quantity 1g

 View more versions of this product

Catalog No. AA1071203

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 16940-92-4
    Formula Weight 441.02
    MDL Number MFCD00010881
    Chemical Name or Material Ammonium hexachloroiridate(IV)
    Grade Premion™

    Specifications

    Physical Form Crystalline Powder
    Formula Weight 441.02
    Health Hazard 3 GHS P Statement
    P234-P264-P270-P301+P312-P406-P501a
    Keep only in original container.
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Store in corrosive resistant container with a resistant inner liner.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H290-H302
    May be corrosive to metals.
    Harmful if swallowed.
    MDL Number MFCD00010881
    Color Red-black
    Health Hazard 1 Warning
    Density 2.86g/mL
    TSCA TSCA
    Sensitivity Hygroscopic
    CAS 16940-92-4
    Quantity 1g
    Concentration Ir 41% min.
    RTECS Number BP5295000
    Chemical Name or Material Ammonium hexachloroiridate(IV)
    Assay 99.994%, (metals basis)
    Grade Premion™

    GHS H Statement
    H290-H302
    May be corrosive to metals.
    Harmful if swallowed.

    GHS P Statement
    P234-P264-P270-P301+P312-P406-P501a
    Keep only in original container.
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Store in corrosive resistant container with a resistant inner liner.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit