Alfa Aesar™ Ammonium hexachloroosmate(IV), Premion™, 99.99% (metals basis), Os 42.8% min

Manufacturer: Alfa Aesar™ 1082302

Quantity 0.5g

 View more versions of this product

Catalog No. AA1082302

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12125-08-5
    Linear Formula (NH4)2OsCl6
    Formula Weight 438.99
    MDL Number MFCD00010883
    Merck Index 14,536
    Chemical Name or Material Ammonium hexachloroosmate(IV)
    Grade Premion™

    Specifications

    Formula Weight 438.99
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Physical Form Powder
    Sensitivity Hygroscopic
    Merck Index 14,536
    Color Brown to black
    Melting Point 170°C (subl.)
    Quantity 0.5g
    Health Hazard 1 Warning
    Density 2.9g/mL
    TSCA TSCA
    Linear Formula (NH4)2OsCl6
    MDL Number MFCD00010883
    CAS 12125-08-5
    Concentration Os 42.8% min.
    Chemical Name or Material Ammonium hexachloroosmate(IV)
    Assay 99.99%, (metals basis)
    Grade Premion™

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit