Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Ammonium hexachlororhodate(III) hydrate, Premion™, 99.99% (metals basis)

Click to view available options
Quantity:
0.25g
1g
5g
15336-18-2
Cl6H12N3Rh
369.723
MFCD00003394
OJNYBYILSUWBHM-UHFFFAOYSA-K
ammonium hexachlororhodate iii,triammonium hexachlororhodate,triammonium hexachlororhodate 3-,epitope id:151523,rhodium iii-ammonium chloride,ammonium hexachloridorhodate 3-,ammonium hexachloridorhodate iii,ammonium hexachlororhodate iii , reagent grade
16211509
CHEBI:60128
triazanium;hexachlororhodium(3-)
[NH4+].[NH4+].[NH4+].Cl[Rh-3](Cl)(Cl)(Cl)(Cl)Cl
15336-18-2
Cl6H12N3Rh
369.723
MFCD00003394
OJNYBYILSUWBHM-UHFFFAOYSA-K
ammonium hexachlororhodate iii,triammonium hexachlororhodate,triammonium hexachlororhodate 3-,epitope id:151523,rhodium iii-ammonium chloride,ammonium hexachloridorhodate 3-,ammonium hexachloridorhodate iii,ammonium hexachlororhodate iii , reagent grade
16211509
CHEBI:60128
triazanium;hexachlororhodium(3-)
[NH4+].[NH4+].[NH4+].Cl[Rh-3](Cl)(Cl)(Cl)(Cl)Cl
Solubility Information Soluble in water.
Formula Weight 369.74 (Anhydrous)
Physical Form Powder
Percent Purity 99.99%
Grade Premion™
Sensitivity Hygroscopic
Color Red
Melting Point (decomposition)
Quantity 5g
Chemical Name or Material Ammonium hexachlororhodate(III) hydrate
Assay (metals basis)
Show More Show Less

GHS H Statement
H318
Causes serious eye damage.

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

EINECSNumber : 239-364-2

TSCA : Yes

Recommended Storage : Ambient temperatures