Alfa Aesar™ Ammonium hexachlororhodate(III) hydrate, Premion™, 99.995% (metals basis)

Manufacturer: Alfa Aesar™ 1084906

Quantity 5g

 View more versions of this product

Catalog No. AA1084906

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 15336-18-2
    Formula Weight 369.74 (Anhydrous)
    MDL Number MFCD00003394
    Chemical Name or Material Ammonium hexachlororhodate(III) hydrate
    Grade Premion™

    Specifications

    Formula Weight 369.74 (Anhydrous)
    Health Hazard 3 GHS P Statement
    P280-P305+P351+P338-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.
    TSCA TSCA
    MDL Number MFCD00003394
    CAS 15336-18-2
    Sensitivity Hygroscopic
    Color Dark red
    Quantity 5g
    Health Hazard 1 Danger
    Physical Form Powder
    Health Hazard 2 GHS H Statement
    H318
    Causes serious eye damage.
    Chemical Name or Material Ammonium hexachlororhodate(III) hydrate
    Assay 99.995%, (metals basis)
    Grade Premion™

    GHS H Statement
    H318
    Causes serious eye damage.

    GHS P Statement
    P280-P305+P351+P338-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit