Alfa Aesar™ Ammonium hexachlorostannate(IV), Puratronic™, 99.999% (metals basis)

Manufacturer: Alfa Aesar™ 4075030

Quantity 250g

 View more versions of this product

Catalog No. AA4075030

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 16960-53-5
    Linear Formula (NH4)2SnCl6
    Formula Weight 367.48
    MDL Number MFCD00036340
    Chemical Name or Material Ammonium hexachlorostannate(IV)
    Grade Puratronic™

    Specifications

    Formula Weight 367.48
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Physical Form Powder
    MDL Number MFCD00036340
    Density 2.4g/mL
    Linear Formula (NH4)2SnCl6
    Quantity 250g
    Solubility Information Soluble in water
    TSCA TSCA
    CAS 16960-53-5
    Color White
    Health Hazard 1 Warning
    Chemical Name or Material Ammonium hexachlorostannate(IV)
    Assay 99.999%, (metals basis)
    Grade Puratronic™

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit