Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Ammonium hexachlorostannate(IV), Puratronic™, 99.999% (metals basis)

Click to view available options
Quantity:
10g
50g
250g
16960-53-5
Cl6H8N2Sn
367.488
MFCD00036340
UDMAVAHFQCVSSE-UHFFFAOYSA-J
ammonium tin chloride,stannic ammonium chloride,ammonium hexachlorostannate,diammonium hexachlorostannate,ammonium hexachlorostannate 2-,diammonium hexachlorostannate 2-,ammonium tin 4+ chloride 2:1:6,diazanium tin 4+ hexachloride,stannate 2-, hexachloro-, diammonium, oc-6-11,stannate 2-, hexachloro-, ammonium 1:2 , oc-6-11
167595
diazanium;tin(4+);hexachloride
[NH4+].[NH4+].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Sn+4]
16960-53-5
Cl6H8N2Sn
367.488
MFCD00036340
UDMAVAHFQCVSSE-UHFFFAOYSA-J
ammonium tin chloride,stannic ammonium chloride,ammonium hexachlorostannate,diammonium hexachlorostannate,ammonium hexachlorostannate 2-,diammonium hexachlorostannate 2-,ammonium tin 4+ chloride 2:1:6,diazanium tin 4+ hexachloride,stannate 2-, hexachloro-, diammonium, oc-6-11,stannate 2-, hexachloro-, ammonium 1:2 , oc-6-11
167595
diazanium;tin(4+);hexachloride
[NH4+].[NH4+].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Sn+4]
Linear Formula (NH4)2SnCl6
Solubility Information Soluble in water
Formula Weight 367.48
Physical Form Powder
Percent Purity 99.999%
Grade Puratronic™
Density 2.4 g/mL
Color White
Melting Point (decomposition)
Quantity 10g
Chemical Name or Material Ammonium hexachlorostannate(IV)
Assay (metals basis)
Show More Show Less

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 241-033-2

TSCA : Yes

Recommended Storage : Ambient temperatures