Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ Ammonium hexachlorostannate(IV), Puratronic™, 99.999% (metals basis)
Click to view available options
Quantity:
10g
50g
250g
Specifications
Linear Formula | (NH4)2SnCl6 |
Solubility Information | Soluble in water |
Formula Weight | 367.48 |
Physical Form | Powder |
Percent Purity | 99.999% |
Grade | Puratronic™ |
Density | 2.4 g/mL |
Color | White |
Melting Point | (decomposition) |
Quantity | 50g |
Show More |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 241-033-2
TSCA : Yes
Recommended Storage : Ambient temperatures