Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Ammonium hexafluoroaluminate, 99.8% (metals basis)

Preparation of aluminum fluoride

Manufacturer:  Alfa Aesar™ 3912814

Quantity 25g

 View more versions of this product

Catalog No. AA3912814

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

7784-19-2
195.09
UN3288
Ammonium hexafluoroaluminate
(NH4)3AlF6
MFCD00042144
14,524

Specifications

7784-19-2
(NH4)3AlF6
MFCD00042144
14,524
1.78g/mL
Odorless
25g
99.8%, (metals basis)
Powder
195.09
UN3288
Freely soluble in water
−80 Mesh
White
Ammonium hexafluoroaluminate

Safety and Handling

SDS & Safety Documents

GHS H Statement
H301-H311-H331
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.

GHS P Statement
P261-P280-P301+P310-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.

RTECSNumber : BD0050000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit