Alfa Aesar™ Ammonium hexafluoroaluminate, 99.8% (metals basis)

Preparation of aluminum fluoride

Manufacturer: Alfa Aesar™ 3912806

Quantity 5g

 View more versions of this product

Catalog No. AA3912806

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 7784-19-2
    Linear Formula (NH4)3AlF6
    Formula Weight 195.09
    MDL Number MFCD00042144
    UN Number UN3288
    Merck Index 14,524
    Chemical Name or Material Ammonium hexafluoroaluminate

    Specifications

    Formula Weight 195.09
    Health Hazard 3 GHS P Statement
    P261-P280-P301+P310-P361-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Color White
    Density 1.78g/mL
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
    RTECS Number BD0050000
    Odor Odorless
    Physical Form Powder
    CAS 7784-19-2
    UN Number UN3288
    Merck Index 14,524
    Solubility Information Freely soluble in water
    Health Hazard 1 Danger
    TSCA TSCA
    Mesh Size −80 Mesh
    Linear Formula (NH4)3AlF6
    MDL Number MFCD00042144
    Quantity 5g
    Health Hazard 2 GHS H Statement
    H301-H311-H331
    Toxic if swallowed.
    Toxic in contact with skin.
    Toxic if inhaled.
    Chemical Name or Material Ammonium hexafluoroaluminate
    Assay 99.8%, (metals basis)

    GHS H Statement
    H301-H311-H331
    Toxic if swallowed.
    Toxic in contact with skin.
    Toxic if inhaled.

    GHS P Statement
    P261-P280-P301+P310-P361-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit