Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium hexafluorosilicate, 99.999%, (trace metal basis), ACROS Organics™

Click to view available options
Quantity:
10g
50g
250g
Packaging:
Glass bottle
16919-19-0
H8F6N2Si
178.15
ITHIMUMYFVCXSL-UHFFFAOYSA-P
ammonium hexafluorosilicate,ammonium fluosilicate,ammonium fluorosilicate,ammonium silicofluoride,cryptohalite,caswell no. 043,diammonium fluosilicate,diammonium fluorosilicate,ammonium silicon fluoride,diammonium hexafluorosilicate
28145
diazanium;hexafluorosilicon(2-)
[NH4+].[NH4+].F[Si-2](F)(F)(F)(F)F
16919-19-0
H8F6N2Si
178.15
ITHIMUMYFVCXSL-UHFFFAOYSA-P
ammonium hexafluorosilicate,ammonium fluosilicate,ammonium fluorosilicate,ammonium silicofluoride,cryptohalite,caswell no. 043,diammonium fluosilicate,diammonium fluorosilicate,ammonium silicon fluoride,diammonium hexafluorosilicate
28145
diazanium;hexafluorosilicon(2-)
[NH4+].[NH4+].F[Si-2](F)(F)(F)(F)F
for ultra trace analysis
Assay Percent Range 99.999%
Linear Formula (NH4)2SiF6
Merck Index 15,524
Solubility Information Solubility in water: 18.6g/100mL (17°C)
Formula Weight 178.15
Physical Form Crystalline Powder and Chunks
Percent Purity 99.999%
Grade Trace Metal Basis
Density 2.0100g/mL
Packaging Glass bottle
Solubility Solubility in water: 18.6 g/100mL (17°C).
Specific Gravity 2.01
Total Trace Metal Impurities 10ppm max.
Color White
Quantity 10g
Chemical Name or Material Ammonium hexafluorosilicate
Show More Show Less

GHS H Statement
Toxic if inhaled.
Toxic in contact with skin.
Toxic if swallowed.

GHS P Statement
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protectio

GHS Signal Word: Danger

EINECSNumber : 240-968-3