Alfa Aesar™ Ammonium hexafluorotitanate, 99.99% (metals basis)

Manufacturer: Alfa Aesar™ 4319236

Quantity 500g

 View more versions of this product

Catalog No. AA4319236

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 16962-40-6
    Molecular Formula (NH4)2TiF6
    Formula Weight 197.95
    MDL Number MFCD00010888
    Chemical Name or Material Ammonium hexafluorotitanate

    Specifications

    Formula Weight 197.95
    Health Hazard 3 GHS P Statement
    P261-P280-P304+P340-P301+P312-P312-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Crystalline
    CAS 16962-40-6
    Quantity 500g
    Molecular Formula (NH4)2TiF6
    MDL Number MFCD00010888
    Color White
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H302-H312-H332
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.
    RTECS Number XR1500000
    TSCA TSCA
    Chemical Name or Material Ammonium hexafluorotitanate
    Assay 99.99%, (metals basis)

    GHS H Statement
    H302-H312-H332
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.

    GHS P Statement
    P261-P280-P304+P340-P301+P312-P312-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit