Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ammonium hydrogen difluoride, 95%, extra pure, ACROS Organics™
Click to view available options
Quantity:
1kg
5kg
Packaging:
Plastic Bottle
Plastic drum
Description
C-X bond formation reagent
Specifications
Additional Information | ammonium fluoride (NH4F): < 5% |
Assay Percent Range | 95% |
Linear Formula | (NH4)HF2 |
Merck Index | 15, 495 |
Solubility Information | Solubility in water: 630g/L (20°C). Other solubilities: very soluble in hot water,soluble in ethanol |
Formula Weight | 57.04 |
Physical Form | Scales or Flakes |
Percent Purity | 95% |
Grade | Extra Pure |
Heavy Metals (as Pb) | 200ppm max. |
Show More |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Toxic if swallowed.
GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
EINECSNumber : 215-676-4
RTECSNumber : BQ9200000
TSCA : TSCA