Alfa Aesar™ Ammonium hydrogen fluoride, tech. 95%

Manufacturer: Alfa Aesar™ 1466036

Quantity 500g

 View more versions of this product

Catalog No. AA1466036

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1341-49-7
    Formula Weight 57.04
    MDL Number MFCD00012018
    UN Number UN1727
    Merck Index 14,498
    Synonym Ammonium bifluoride
    Chemical Name or Material Ammonium hydrogen fluoride
    Grade Technical Grade

    Specifications

    Formula Weight 57.04
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H300-H314-H318
    Fatal if swallowed.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Odor Ammonia-like
    Sensitivity Hygroscopic
    Density 1.500g/mL
    RTECS Number BQ9200000
    Physical Form Flakes
    CAS 1341-49-7
    UN Number UN1727
    Synonym Ammonium bifluoride
    pH 2 to 3
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMMONIUM HYDROGENDIFLUORIDE, SOLID
    TSCA TSCA
    MDL Number MFCD00012018
    Merck Index 14,498
    Boiling Point 238°C
    Melting Point 120° to 122°C
    Quantity 500g
    Chemical Name or Material Ammonium hydrogen fluoride
    Assay 95%
    Grade Technical Grade

    GHS H Statement
    H300-H314-H318
    Fatal if swallowed.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit