Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium hydrogen sulfate, extra pure, ACROS Organics™

Click to view available options
Quantity:
1kg
2.5kg
7803-63-6
H5NO4S
115.11
BIGPRXCJEDHCLP-UHFFFAOYSA-N
ammonium bisulfate,ammonium hydrogensulphate,sulfuric acid, monoammonium salt,unii-6218r7mbzb,sulfuric acid, ammonium salt 1:1,azanium hydrogen sulfate,ammonia sulfate,acmc-1bcew,ammonium hydrogen sulphate,nh4hso4
24655
azanium;hydrogen sulfate
[NH4+].OS(=O)(=O)[O-]
7803-63-6
H5NO4S
115.11
BIGPRXCJEDHCLP-UHFFFAOYSA-N
ammonium bisulfate,ammonium hydrogensulphate,sulfuric acid, monoammonium salt,unii-6218r7mbzb,sulfuric acid, ammonium salt 1:1,azanium hydrogen sulfate,ammonia sulfate,acmc-1bcew,ammonium hydrogen sulphate,nh4hso4
24655
azanium;hydrogen sulfate
[NH4+].OS(=O)(=O)[O-]
Assay Percent Range Extra pure
Linear Formula NH4HSO4
Merck Index 15,497
Solubility Information Solubility in water: 1000g/L (20°C). Other solubilities: practically insoluble in alcohol,acetone and,pyridine
Formula Weight 115.11
Grade Extra Pure
Chloride 0.005% max
Insoluble Matter 0.05% max
Packaging Glass bottle
Phosphate 0.003% max
Solubility Solubility in water: 1000g/L (20°C). Other solubilities: practically insoluble in alcohol, acetone and, pyridine
Boiling Point 350.0°C
Melting Point 147.0°C
Quantity 1kg
Chemical Name or Material Ammonium hydrogen sulfate
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

EINECSNumber : 232-265-5