Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Hydroxide, 0.2, BAKER INSTRA-ANALYZED™ Plus, J.T.Baker™

Click to view available options
Quantity:
500mL
4L
Packaging:
Poly Bottle
Pressure Bottom Poly Bottle
1336-21-6
H5NO
35.046
VHUUQVKOLVNVRT-UHFFFAOYSA-N
ammonium hydroxide,ammonia water,ammonia aqueous,aquammonia,aqua ammonia,ammonia, aqua,household ammonia,ammonium hydroxide solution,ammonia, monohydrate,ammonium hydroxide nh4 oh
14923
CHEBI:18219
azanium;hydroxide
[NH4+].[OH-]
1336-21-6
H5NO
35.046
VHUUQVKOLVNVRT-UHFFFAOYSA-N
ammonium hydroxide,ammonia water,ammonia aqueous,aquammonia,aqua ammonia,ammonia, aqua,household ammonia,ammonium hydroxide solution,ammonia, monohydrate,ammonium hydroxide nh4 oh
14923
CHEBI:18219
azanium;hydroxide
[NH4+].[OH-]

Acids for critical trace metal analysis; Quality is assured for up to 64 trace metals tested to very low parts-per-billion (ppb) levels

    For use in ICP (Inductively Coupled Plasma Spectrometry), Trace Metal Analysis; For Laboratory, Research, or Manufacturing Use
Quantity 500mL
Formula Weight 35.05
Percent Purity 20%
Grade BAKER INSTRA-ANALYZED™ Plus
Packaging Poly Bottle
Specific Gravity 1L = 0.93kg
Chemical Name or Material Ammonium Hydroxide