Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Hydroxide, 0.5 N Solution, For Phenols, APHA, Spectrum™

Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Poly-Coated Glass Bottle
1336-21-6,7732-18-5,7664-41-7
1336-21-6,7732-18-5,7664-41-7
Concentration 0.5 N ± 0.02 N
Grade Chemical Solution
Packaging Poly-Coated Glass Bottle
Quantity 500 mL

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Material Safety Data Sheet (MSDS) prior to use.