Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Hydroxide, 1% (1+99), Solution, Spectrum™

Click to view available options
Quantity:
500 mL
Packaging:
Poly Bottle
CAS 1336-21-6,7732-18-5
Grade Chemical Solution
Packaging Poly Bottle
Quantity 500 mL

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.