Ammonium Hydroxide, 16.7% (v/v) Aqueous Solution, Electronic Grade, Ricca Chemical

Manufacturer: Ricca Chemical CompanySupplier Diversity Partner 6325

Packaging Cubitainer™
Quantity 20L

 View more versions of this product

Catalog No. 632-5

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7732-18-5,1336-21-6
    CAS Min % 83.0,16.0
    CAS Max % 84.0,17.0
    Physical Form Liquid
    Chemical Name or Material Ammonium Hydroxide
    Grade Electronic Grade

    Specifications

    Physical Form Liquid
    Color Clear colorless
    Quantity 20L
    Concentration or Composition Notes 16.7% (v/v)
    Solvent or Matrix Aqueous Solution
    CAS 1336-21-6,7732-18-5
    CAS Max % 17.0,84.0
    CAS Min % 83.0,16.0
    Packaging Cubitainer™
    Chemical Name or Material Ammonium Hydroxide
    Grade Electronic Grade

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit