Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Hydroxide, 20% (v/v; 1+4) Solution, For Hardness, ASTM, Spectrum™

Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Poly Bottle
1336-21-6,7732-18-5
1336-21-6,7732-18-5
Grade Chemical Solution
Packaging Poly Bottle
Quantity 4 L

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.