Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Hydroxide, 25% (1+3) Solution, Spectrum™

Click to view available options
Quantity:
500 mL
1 L
20 L
Packaging:
Amber Glass Bottle
Cubitainer
1336-21-6,7732-18-5
1336-21-6,7732-18-5
CAS Max % 1
Grade Chemical Solution
Packaging Amber Glass Bottle
Quantity 1 L

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.